ENA333557054 Enamine

4-(4H-1,2,4-triazol-3-yl)phenol

  • MDL number MFCD19686229
  • Cas Number : 1334149-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77930
Availability details for
Quantity
  • Availability
  • Add to Cart