ENA333557058 Enamine

3-acetamido-4-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD19686235
  • Cas Number : 160233-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79102
Availability details for
Quantity
  • Availability
  • Add to Cart