ENA333557068 Enamine

bis(N-[2-(dimethylamino)ethyl]guanidine); sulfuric acid

  • MDL number MFCD19686245
  • Cas Number : 905300-72-3
  • Molecular Weight : 358.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2H2SO4
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-80508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80508
Availability details for
Quantity
  • Availability
  • Add to Cart