ENA333557075 Enamine

2-chloroprop-2-en-1-amine hydrochloride

  • MDL number MFCD19686253
  • Cas Number : 100516-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-213
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80822
Availability details for
Quantity
  • Availability
  • Add to Cart