ENA333557079 Enamine

methyl({[1-(pyridazin-3-yl)pyrrolidin-2-yl]methyl})amine dihydrobromide

  • MDL number MFCD19686258
  • Cas Number : 1334149-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80848
Availability details for
Quantity
  • Availability
  • Add to Cart