ENA333557086 Enamine

2-amino-4-fluoro-5-(4-methyl-1H-pyrazol-1-yl)benzoic acid hydrochloride

  • MDL number MFCD19686266
  • Cas Number : 1334148-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80902
Availability details for
Quantity
  • Availability
  • Add to Cart