ENA333557089 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-(5-methyl-1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD19686272
  • Cas Number : 1334148-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80981
Availability details for
Quantity
  • Availability
  • Add to Cart