ENA333557091 Enamine

4-(2-aminoethyl)-5-cyclopropyl-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD19686274
  • Cas Number : 1334147-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80994

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80994
Availability details for
Quantity
  • Availability
  • Add to Cart