ENA333557100 Enamine

thieno[3,2-b]furan-5-carboxylic acid

  • MDL number MFCD19686287
  • Cas Number : 1334148-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81532
Availability details for
Quantity
  • Availability
  • Add to Cart