ENA333557112 Enamine

2,2,2-trifluoroethyl N-(2-carbamoyl-4-fluoro-5-methylphenyl)carbamate

  • MDL number MFCD19686299
  • Cas Number : 1334148-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81593
Availability details for
Quantity
  • Availability
  • Add to Cart