ENA333557113 Enamine

2-amino-5-fluoro-4-methylbenzamide

  • MDL number MFCD19686300
  • Cas Number : 1334149-42-6
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-fluoro-4-methylbenzamide
Inchi Code 1S/C8H9FN2O/c1-4-2-7(10)5(8(11)12)3-6(4)9/h2-3H,10H2,1H3,(H2,11,12)
Inchi Key HLZUHAKHLQHOJH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-81594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81594
Availability details for
Quantity
  • Availability
  • Add to Cart