ENA333557117 Enamine

4-(4-fluorophenyl)-2-methylpyrrolidine

  • MDL number MFCD19686307
  • Cas Number : 1334148-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81605
Availability details for
Quantity
  • Availability
  • Add to Cart