ENA333557124 Enamine

1-methyl-6-oxo-2-(thiophen-2-yl)piperidine-3-carboxylic acid

  • MDL number MFCD07378149
  • Cas Number : 1334148-77-4
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-81636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81636
Availability details for
Quantity
  • Availability
  • Add to Cart