ENA333557125 Enamine

5-(hydroxymethyl)-1-methyl-6-[4-(trifluoromethyl)phenyl]piperidin-2-one

  • MDL number MFCD19686315
  • Cas Number : 1334146-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81638
Availability details for
Quantity
  • Availability
  • Add to Cart