ENA333557134 Enamine

2-(1-ethoxyethyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD19686324
  • Cas Number : 1334146-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81677
Availability details for
Quantity
  • Availability
  • Add to Cart