ENA333557137 Enamine

4-[(4-aminophenyl)amino]butanoic acid

  • MDL number MFCD19686327
  • Cas Number : 86147-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81682
Availability details for
Quantity
  • Availability
  • Add to Cart