ENA333557153 Enamine

2-chloro-N-(2-oxo-2-phenylethyl)propanamide

  • MDL number MFCD19686345
  • Cas Number : 1334149-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-81774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81774
Availability details for
Quantity
  • Availability
  • Add to Cart