ENA333557154 Enamine

1-(5-chloro-2-methoxyphenyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD19686346
  • Cas Number : 1334148-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81778
Availability details for
Quantity
  • Availability
  • Add to Cart