ENA333557158 Enamine

5-(3-chlorophenyl)-4-(hydroxymethyl)-1-methylpyrrolidin-2-one

  • MDL number MFCD19686352
  • Cas Number : 1334149-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81796
Availability details for
Quantity
  • Availability
  • Add to Cart