ENA333557196 Enamine

sodium 2-[hydroxy(phenyl)methyl]benzoate

  • MDL number MFCD19686390
  • Cas Number : 1334148-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-81859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81859
Availability details for
Quantity
  • Availability
  • Add to Cart