ENA333557197 Enamine

N-ethylcyclobutanamine hydrochloride

  • MDL number MFCD19686391
  • Cas Number : 1334146-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81860

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81860
Availability details for
Quantity
  • Availability
  • Add to Cart