ENA333557202 Enamine

4-(4-fluorophenyl)-2-(pyrrolidin-2-yl)-1H-imidazole

  • MDL number MFCD12105656
  • Cas Number : 1155056-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81890

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81890
Availability details for
Quantity
  • Availability
  • Add to Cart