ENA333557203 Enamine

4-(2-fluorophenyl)-2-(pyrrolidin-2-yl)-1H-imidazole

  • MDL number MFCD12105640
  • Cas Number : 1153102-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81891
Availability details for
Quantity
  • Availability
  • Add to Cart