ENA333557204 Enamine

2,2,5-trimethylhexan-3-amine hydrochloride

  • MDL number MFCD19686400
  • Cas Number : 1334149-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81904
Availability details for
Quantity
  • Availability
  • Add to Cart