ENA333557205 Enamine

4-(3-chloropropyl)-1H-pyrazole hydrochloride

  • MDL number MFCD19686401
  • Cas Number : 178620-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81911
Availability details for
Quantity
  • Availability
  • Add to Cart