ENA333557208 Enamine

N-[2-methyl-1-(5-methyl-4H-1,2,4-triazol-3-yl)propyl]benzamide

  • MDL number MFCD19686404
  • Cas Number : 1334147-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81915
Availability details for
Quantity
  • Availability
  • Add to Cart