ENA333557209 Enamine

4-(4-methoxyphenyl)-2-methylpyrrolidine

  • MDL number MFCD19686406
  • Cas Number : 91563-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81929
Availability details for
Quantity
  • Availability
  • Add to Cart