ENA333557223 Enamine

N-(2-phenoxyethyl)piperidin-4-amine dihydrochloride

  • MDL number MFCD19686420
  • Cas Number : 1334147-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-81970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81970
Availability details for
Quantity
  • Availability
  • Add to Cart