ENA333557226 Enamine

3-(2-ethyl-1H-imidazol-1-yl)piperidine dihydrochloride

  • MDL number MFCD19686423
  • Cas Number : 1334149-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81994
Availability details for
Quantity
  • Availability
  • Add to Cart