ENA333557236 Enamine

methyl 3-amino-5-chlorobenzoate

  • MDL number MFCD11053764
  • Cas Number : 21961-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77882
Availability details for
Quantity
  • Availability
  • Add to Cart