ENA333557238 Enamine

1-(piperazin-1-yl)phthalazine dihydrochloride

  • MDL number MFCD19982346
  • Cas Number : 1333898-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 309...311
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77919

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77919
Availability details for
Quantity
  • Availability
  • Add to Cart