ENA333557240 Enamine

2-chloro-1-(5-methoxy-2-methyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD03848172
  • Cas Number : 843628-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77968

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77968
Availability details for
Quantity
  • Availability
  • Add to Cart