ENA333557245 Enamine

4-(chloromethyl)-2-fluoro-1-(2-fluorophenoxy)benzene

  • MDL number MFCD11190677
  • Cas Number : 1039960-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78279
Availability details for
Quantity
  • Availability
  • Add to Cart