ENA333557246 Enamine

ethyl 3-hydroxy-2,3-dimethylbutanoate

  • MDL number MFCD19686230
  • Cas Number : 34849-39-3
  • Molecular Weight : 160.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78341
Availability details for
Quantity
  • Availability
  • Add to Cart