ENA333557267 Enamine

2-(dimethyl-1,3-thiazol-2-yl)propan-2-amine

  • MDL number MFCD11587499
  • Cas Number : 1082478-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79190
Availability details for
Quantity
  • Availability
  • Add to Cart