ENA333557270 Enamine

pentafluorophenyl 2-chloroacetate

  • MDL number MFCD19686239
  • Cas Number : 240487-99-4
  • Molecular Weight : 260.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3,4,5,6-pentafluorophenyl chloroacetate
Inchi Code 1S/C8H2ClF5O2/c9-1-2(15)16-8-6(13)4(11)3(10)5(12)7(8)14/h1H2
Inchi Key VVKNQIGLUQLDPD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79695
Availability details for
Quantity
  • Availability
  • Add to Cart