ENA333557275 Enamine

methyl 3-(5-amino-1,3,4-thiadiazol-2-yl)propanoate

  • MDL number MFCD14543230
  • Cas Number : 227958-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79763
Availability details for
Quantity
  • Availability
  • Add to Cart