ENA333557280 Enamine

5-{[(2-fluorophenyl)methyl]amino}-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD06752574
  • Cas Number : 876897-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79853
Availability details for
Quantity
  • Availability
  • Add to Cart