ENA333557289 Enamine

benzyl[2-(methylamino)ethyl]amine

  • MDL number MFCD09037852
  • Cas Number : 56904-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50645
Availability details for
Quantity
  • Availability
  • Add to Cart