ENA333557300 Enamine

5-phenylpent-4-enoic acid

  • MDL number MFCD01926594
  • Cas Number : 28525-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80430

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80430
Availability details for
Quantity
  • Availability
  • Add to Cart