ENA333557310 Enamine

6-fluoro-3-methyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD13196326
  • Cas Number : 863119-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80539
Availability details for
Quantity
  • Availability
  • Add to Cart