ENA333557311 Enamine

methyl 2-methyl-3-oxopentanoate

  • MDL number MFCD00184703
  • Cas Number : 17422-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80573
Availability details for
Quantity
  • Availability
  • Add to Cart