ENA333557314 Enamine

2-sulfanyl-1,3-benzoxazole-6-carboxylic acid

  • MDL number MFCD09259279
  • Cas Number : 108085-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 295...297
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80628

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80628
Availability details for
Quantity
  • Availability
  • Add to Cart