ENA333557320 Enamine

N-[3-(aminomethyl)phenyl]-4-chlorobenzene-1-sulfonamide

  • MDL number MFCD19124851
  • Cas Number : 1274702-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-80656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80656
Availability details for
Quantity
  • Availability
  • Add to Cart