ENA333557323 Enamine

3-[(4-cyanophenyl)amino]benzoic acid

  • MDL number MFCD11550349
  • Cas Number : 1098383-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80685
Availability details for
Quantity
  • Availability
  • Add to Cart