ENA333557324 Enamine

2-(chloromethyl)-6-methylpyridine hydrochloride

  • MDL number MFCD01691969
  • Cas Number : 3099-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80687
Availability details for
Quantity
  • Availability
  • Add to Cart