ENA333557327 Enamine

ethyl 2-amino-5-bromo-3-chlorobenzoate

  • MDL number MFCD12159924
  • Cas Number : 1032668-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80721

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80721
Availability details for
Quantity
  • Availability
  • Add to Cart