ENA333557333 Enamine

3-(2,2,2-trifluoroethoxy)propan-1-ol

  • MDL number MFCD14650970
  • Cas Number : 215929-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53971
Availability details for
Quantity
  • Availability
  • Add to Cart