ENA333557358 Enamine

methyl[1-(methylsulfanyl)propan-2-yl]amine

  • MDL number MFCD16850260
  • Cas Number : 1248277-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80834
Availability details for
Quantity
  • Availability
  • Add to Cart