ENA333557359 Enamine

N-[(4-aminophenyl)methyl]-N-cyclopropylacetamide

  • MDL number MFCD12175073
  • Cas Number : 1156263-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80835
Availability details for
Quantity
  • Availability
  • Add to Cart